Learn More
Medchemexpress LLC HY-P1026 1mg Medchemexpress, NFAT Inhibitor CAS:249537-73-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP10261MG
Encompass_Preferred
Medchemexpress, HY-P1026 1mg NFAT Inhibitor CAS:249537-73-3 NFAT Inhibitor (VIVIT peptide; 11R-VIVIT) is a selective inhibitor of calcineurin-mediated dephosphorylation of nuclear factor of activated T cells (NFAT). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4135