Learn More
TARGETMOL CHEMICALS INC NFAT Inhibitor 5MG
SDP
Catalog No. 502879630
Encompass_Preferred
Also available in 1 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. NFAT Inhibitor (VIVIT peptide) is a cell-permeable compound that selectively inhibits the calcineurin-mediated dephosphorylation of NFAT. Purity 98.26%
Catalog No. 50-287-9630 Supplier TARGETMOL CHEMICALS INC Supplier No. TP10155MG
Also available in 1 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. NFAT Inhibitor (VIVIT peptide) is a cell-permeable compound that selectively inhibits the calcineurin-mediated dephosphorylation of NFAT. Purity 98.26%