Learn More
Cayman Chemical NFAT InhIbItrcel-permeabl 1mg
SDP
Catalog No. 502909154
Encompass_Preferred
A cell-permeable form of NFAT inhibitor; inhibits IL-12p40 and NO production in LPS- and IFN-γ-induced mouse BMDMs at 50 or 100 µM; prevents increases in kidney weight and urine albumin levels in a db/db mouse model of diabetic nephropathy at 1 mg/kg; improves fracture healing in a rat model of ovariectomy-induced osteoporosis at 100 UG/kg for 16 weeks post-fracture; decreases disease severity in IL-10 knockout mice in a model of piroxicam-induced colitis at 1 mg/kg
Catalog No. 50-290-9154 Supplier Cayman Chemical Supplier No. 360661 MG
A cell-permeable form of NFAT inhibitor; inhibits IL-12p40 and NO production in LPS- and IFN-γ-induced mouse BMDMs at 50 or 100 µM; prevents increases in kidney weight and urine albumin levels in a db/db mouse model of diabetic nephropathy at 1 mg/kg; improves fracture healing in a rat model of ovariectomy-induced osteoporosis at 100 UG/kg for 16 weeks post-fracture; decreases disease severity in IL-10 knockout mice in a model of piroxicam-induced colitis at 1 mg/kg