Learn More
BPS Bioscience Inc NFAT Reporter (eGFP) Jurkat Recombinant Cell Line, 2 vials
SDP
Supplier:  BPS Bioscience Inc 78384
Encompass
The NFAT Reporter (eGFP) Jurkat Recombinant Cell Line is designed for scientific applications focusing on T-cell activation monitoring. Engineered from Jurkat T-cells, these cells express an enhanced Green Fluorescent Protein (eGFP) under the regulation of NFAT response elements, providing a visual means to assess NFAT signaling pathway activation. This product is ideal for use with immune checkpoint inhibitors and for co-expression studies with modulators like PD1. In immune responses, NFAT plays a critical role, with its activity linked to calcium influx and cytokine gene induction (e.g., IL-2, IL-3, IL-4, TNF-alpha).
Catalog No. NC2890886