Learn More
Medchemexpress LLC HY-112085 10mg Medchemexpress, N-Bis(2-hydroxypropyl)nitrosamine CAS:53609-64-6 Purity:>98%
SDP
Catalog No. 501969018
Encompass_Preferred
Medchemexpress, HY-112778 10mg NFAT Transcription Factor Regulator-1 CAS:245747-71-1 NFAT Transcription Factor Regulator-1 is an IL-2 synthesis inhibitor with an IC50 of 182 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9018 Supplier Medchemexpress LLC Supplier No. HY11208510MG
Medchemexpress, HY-112778 10mg NFAT Transcription Factor Regulator-1 CAS:245747-71-1 NFAT Transcription Factor Regulator-1 is an IL-2 synthesis inhibitor with an IC50 of 182 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.