Learn More
ABclonal Technology NFAT2 Rabbit mAb
SDP
Catalog No. 5000006758
Encompass_Preferred
The product of this gene is a component of the nuclear factor of activated T cells DNA-binding transcription complex This complex consists of at least two components a preexisting cytosolic component that translocates to the nucleus upon T cell receptor (TCR) stimulation and an inducible nuclear component Proteins belonging to this family of transcription factors play a central role in inducible gene transcription during immune response The product of this gene is an inducible nuclear component It functions as a major molecular target for the immunosuppressive drugs such as cyclosporin A Multiple alternatively spliced transcript variants encoding distinct isoforms have been identified for this gene Different isoforms of this protein may regulate inducible expression of different cytokine genes
Catalog No. 50-000-06758 Supplier ABclonal Technology Supplier No. A2348520UL
The product of this gene is a component of the nuclear factor of activated T cells DNA-binding transcription complex This complex consists of at least two components a preexisting cytosolic component that translocates to the nucleus upon T cell receptor (TCR) stimulation and an inducible nuclear component Proteins belonging to this family of transcription factors play a central role in inducible gene transcription during immune response The product of this gene is an inducible nuclear component It functions as a major molecular target for the immunosuppressive drugs such as cyclosporin A Multiple alternatively spliced transcript variants encoding distinct isoforms have been identified for this gene Different isoforms of this protein may regulate inducible expression of different cytokine genes