Learn More
Aviva Systems Biology Corp NFATC1 Antibody - N-terminal region (ARP38742_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38742_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NFATC1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human NFATC1. Alternate names: MGC138448, NF-ATC, NFAT2, NFATcApplicationWB, IHC. Species reactivity: Guinea Pig, Human, Mouse, Rat
Catalog No. 50-014-56778