Learn More
Aviva Systems Biology Corp NFATC1 Antibody - C-terminal region (ARP39978_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39978_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NFATC1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human NFATC1. Alternate names: MGC138448, NF-ATC, NFAT2, NFATcApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-284-844