Learn More
Cayman Chemical NFATc1 InhIbItr A04 100mg
SDP
Catalog No. 502944389
Encompass_Preferred
An NFATc1 inhibitor; inhibits RANKL-induced nuclear translocation of NFATc1 in RAW 264.7 cells at 1.5 or 2 µM; reduces RANKL-induced differentiation of RAW 264.7 cells into osteoclasts (IC50 = 1.57 µM)
Catalog No. 50-294-4389 Supplier Cayman Chemical Supplier No. 41521100 MG
An NFATc1 inhibitor; inhibits RANKL-induced nuclear translocation of NFATc1 in RAW 264.7 cells at 1.5 or 2 µM; reduces RANKL-induced differentiation of RAW 264.7 cells into osteoclasts (IC50 = 1.57 µM)