Learn More
Aviva Systems Biology Corp NFATC2 Antibody - C-terminal region (ARP38988_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38988_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NFATC2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human NFATC2. Alternate names: NFAT1, NFATPApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-284-770