Learn More
AdipoGen NFATc2 h pAb IG-209
SDP
Supplier:  AdipoGen AG25T0111C020
Encompass_Preferred
Polyclonal Antibody. Recognizes human NFATc2. Source Rabbit. Applications ICC, IHC, IP, WB. Liquid. In PBS containing 1mg/ml BSA and 0.02% sodium azide. NFATc2 plays a role in the inducible expression of cytokine genes in T cells, especially in the induction of IL-2, IL-3, IL-4, TNF-alpha or GM-CSF. It promotes invasive migration through the activation of GPC6 expression and the WNT5A signaling pathway.
Catalog No. 50-256-3162