Learn More
Aviva Systems Biology Corp NFATC3 Antibody - N-terminal region (P100975_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp P100975_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NFATC3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human NFATC3. Alternate names: NFAT4, NFATXApplicationIHC, WB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Rat
Catalog No. 50-014-62811