Learn More
Aviva Systems Biology Corp NFATC4 Antibody - middle region (ARP32715_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32715_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against NFATC4. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human NFATC4. Alternate names: NFAT3, NF-ATc4ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-54721