Learn More
Aviva Systems Biology Corp NFATC4 Antibody - N-terminal region (ARP32040_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32040_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NFATC4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human NFATC4. Alternate names: NF-ATc4, NFAT3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54433