Learn More
Cayman Chemical Nefazodone-d6 (hydrochloride), Purity: ≥99% deuterated forms (d1-d6), CAS: 1330236-06-0, 5 mg
SDP
Catalog No. 502946476
Encompass_Preferred
An internal standard for the quantification of nefazodone by GC- or LC-MS
Catalog No. 50-294-6476 Supplier Cayman Chemical Supplier No. 342645 MG
An internal standard for the quantification of nefazodone by GC- or LC-MS