Learn More
Aviva Systems Biology Corp NFE2L1 Antibody - middle region (ARP38270_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38270_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NFE2L1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human NFE2L1. Alternate names: FLJ00380, LCR-F1, NRF1, TCF11ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-56586