Learn More
Aviva Systems Biology Corp NFE2L3 Antibody - middle region (ARP33727_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33727_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NFE2L3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human NFE2L3. Alternate names: NRF3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-52923