Learn More
CPC Scientific MCA-Arg-Pro-Lys-Pro-Tyr-Ala-Nva-Trp-Met-Lys(DNP)-NH2 (trifluoroacetate salt) 1MG
SDP
Catalog No. 502109798
Encompass
Substrate that is hydrolyzed 60 times more rapidly by stromelysin 1 (MMP-3) (kcat/Km= 59400 M-1s-1) than by interstitial collagenase (MMP-1). However, it presents little discrimination between MMP-3, gelatinase A (MMP-2)(kcat/Km= 5400 M-1s-1), and gelatinase B (MMP-9)(kcat/Km= 55300 M-1s-1).SEQUENCE: MCA-Arg-Pro-Lys-Pro-Tyr-Ala-Nva-Trp-Met-Lys(DNP)-NH2 (trifluoroacetate salt)ONE-LETTER SEQUENCE: Mca-RPKPYA-Nva-WM-Lys(Dnp)-NH2MOLECULAR FORMULA: C79H105N19O19S1MOLECULAR WEIGHT: 1656.89STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [158584-08-8]SYNONYMS: Mca-Arg-Pro-Lys-Pro-Tyr-Ala-Nva-Trp-Met-Lys(Dnp)-NHRESEARCH AREA: CancerREFERENCES:H. Nagase, et al., J. Biol. Chem., 269, 20952 (1994)SKU(s): MMPS-024A, MMPS-024B, MMPS-024C
Catalog No. 50-210-9798 Supplier CPC Scientific Supplier No. MMPS024B
Add to Cart
Add to Cart
Substrate that is hydrolyzed 60 times more rapidly by stromelysin 1 (MMP-3) (kcat/Km= 59400 M-1s-1) than by interstitial collagenase (MMP-1). However, it presents little discrimination between MMP-3, gelatinase A (MMP-2)(kcat/Km= 5400 M-1s-1), and gelatinase B (MMP-9)(kcat/Km= 55300 M-1s-1).SEQUENCE: MCA-Arg-Pro-Lys-Pro-Tyr-Ala-Nva-Trp-Met-Lys(DNP)-NH2 (trifluoroacetate salt)ONE-LETTER SEQUENCE: Mca-RPKPYA-Nva-WM-Lys(Dnp)-NH2MOLECULAR FORMULA: C79H105N19O19S1MOLECULAR WEIGHT: 1656.89STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [158584-08-8]SYNONYMS: Mca-Arg-Pro-Lys-Pro-Tyr-Ala-Nva-Trp-Met-Lys(Dnp)-NHRESEARCH AREA: CancerREFERENCES:H. Nagase, et al., J. Biol. Chem., 269, 20952 (1994)SKU(s): MMPS-024A, MMPS-024B, MMPS-024C