Learn More
CPC Scientific MCA-Arg-Pro-Lys-Pro-Gln-OH (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific MMPS025A
Encompass
ONE-LETTER SEQUENCE: Mca-RPKPQMOLECULAR FORMULA: C39H56N10O11MOLECULAR WEIGHT:840.92STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [396718-26-6], RESEARCH AREA: CancerREFERENCES: H. Nagase, et al., J. Biol. Chem., 269, 20952 (1994)
Catalog No. 50-210-9800