Learn More
CPC Scientific MCA-Arg-Pro-Lys-Pro-Val-Glu-Nva-Trp-Arg-Lys(DNP)-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific MMPS026B
Encompass
ONE-LETTER SEQUENCE: Mca-RPKPVE-Nva-WRK(DNP)-NH2MOLECULAR FORMULA: C78H110N22O20MOLECULAR WEIGHT:1675.9STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [158584-09-9], RESEARCH AREA: CancerREFERENCES: H. Nagase, et al., J. Biol. Chem., 269, 20952 (1994)
Catalog No. 50-210-9803