Learn More
Aviva Systems Biology Corp NFIC Antibody - C-terminal region (P100819_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp P100819_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against NFIon Channel. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human NFIC. Alternate names: CTF, NFI, CTF5, NF-IApplicationWB. Species reactivity: Human
Catalog No. 50-014-62728