Learn More
Aviva Systems Biology Corp NFIL3 Antibody - N-terminal region (ARP38613_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38613_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against NFIL3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human NFIL3. Alternate names: E4BP4, IL3BP1, NFIL3A, NF-IL3AApplicationWB, IHC. Species reactivity: Cow, Dog, Human, Pig, Sheep
Catalog No. 50-014-56729