Learn More
Aviva Systems Biology Corp NFIX Antibody - middle region (ARP32717_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32717_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NFIX. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human NFIX. Alternate names: NF1AApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-54723