Learn More
Enzo Life Sciences NF-kappaB p50 subunit (human) (35-381), (recombinant) (GST-tag) (50 µg)
SDP
Supplier:  Enzo Life Sciences BMLUW99800050
Encompass_Preferred
Purity: ≥95% (SDS-PAGE). Formulation: Liquid. In 50mM HEPES, pH 7.5, 50mM NaCl, 1mM DTT. Source: Produced in E. coli. Long Term Storage: -80°C.
Catalog No. 50-201-2342