Learn More
Santa Cruz Biotechnology NFKB P65 (F-6) MOUSE MONO 200U
Supplier:  Santa Cruz Biotechnology SC8008
Encompass
NFKB p65 (F-6),mouse monoclonal IgG1; 200 ug/ml,raised against amino acids 1-286 mapping at the N-terminus of NFB p65 of human origin,recommended for detection of NFKB p65 of m, r, h origin by WB, IP, IHC(P) and FCM,conjugate for IP studies
Catalog No. NC9775128