Learn More
Enzo Life Sciences NF-kappaB p65 subunit (human) (12-317), (recombinant) (GST-tag) (50 µg)
SDP
Supplier:  Enzo Life Sciences BMLUW99900050
Encompass_Preferred
Purity: ≥85% (SDS-PAGE). Formulation: Liquid. In 50mM HEPES, pH 7.5, containing 50mM sodium chloride and 1mM DTT. Source: Produced in E. coli BL21 (λDE3). Recombinant human NF-κB p65 subunit (12-317) is fused to a GST-tag. Long Term Storage: -80°C.
Catalog No. 50-201-2344