Learn More
Aviva Systems Biology Corp NFKB1 Antibody - N-terminal region (ARP38413_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38413_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NFKB1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human NFKB1. Alternate names: p50, KBF1, p105, EBP-1, NFKB-p50, NFkappaB, NF-kappaB, NFKB-p105, NF-kappa-BApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56643