Learn More
Aviva Systems Biology Corp NFKB1 Antibody - middle region (AVARP09048_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP09048_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NFKB1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human NFKB1. Alternate names: DKFZp686C01211, EBP-1, KBF1, MGC54151, NF-kappa-B, NFKB-p105, NFKB-p50, p50, p105, NFkappaB, NF-kappaBApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-62587