Learn More
Aviva Systems Biology Corp NFKBIA Antibody - middle region (AVARP09044_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP09044_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NFKBIA. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human NFKBIA. Alternate names: IKBA, MAD-3, NFKBIApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rat, Sheep
Catalog No. 50-014-62583