Learn More
Aviva Systems Biology Corp NFKBIB Antibody - C-terminal region (ARP34504_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34504_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against NFKBIB. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human NFKBIB. Alternate names: IKBB, TRIP9ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55424