Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC NFKBIE/IKB epsilon protein, human (His tag) | 50UG
SDP

Catalog No. 5000175391
Encompass_Preferred

Recombinant human NFKBIE (IκBε) protein with a His tag, expressed in E. coli and supplied lyophilized for in vitro biochemical and cell-signaling studies of NF-κB regulation. Lyophilized material is provided from a buffered formulation and is stable when stored at recommended temperatures.

  • Recombinant human NFKB inhibitor epsilon with a His tag
  • Expressed in E. coli and purified for research use
  • Sequence length 395 amino acids and approximate mass 57 kDa
  • Lyophilized from 50 mM Tris-HCl, 200 mM NaCl, 200 mM arginine, pH 8.0, 10% glycerol
  • Store frozen; stable at -20°C for long term and at 4°C for short term after reconstitution

Catalog No. 50-001-75391 Supplier Medchemexpress LLC Supplier No. HYP70123050UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human NFKBIE (IκBε) protein with a His tag, expressed in E. coli and supplied lyophilized for in vitro biochemical and cell-signaling studies of NF-κB regulation. Lyophilized material is provided from a buffered formulation and is stable when stored at recommended temperatures.

  • Recombinant human NFKB inhibitor epsilon with a His tag
  • Expressed in E. coli and purified for research use
  • Sequence length 395 amino acids and approximate mass 57 kDa
  • Lyophilized from 50 mM Tris-HCl, 200 mM NaCl, 200 mM arginine, pH 8.0, 10% glycerol
  • Store frozen; stable at -20°C for long term and at 4°C for short term after reconstitution

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.