Learn More
Aviva Systems Biology Corp NFKBIL1 Antibody - N-terminal region (ARP38540_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38540_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NFKBIL1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human NFKBIL1. Alternate names: IKBL, LST1, NFKBILApplicationWB. Species reactivity: Cow, Dog, Goat, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-56694