Learn More
Aviva Systems Biology Corp Nfkbil1 Antibody - C-terminal region (ARP38541_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38541_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Nfkbil1. It was validated on Western Blot. . Alternate names: Def-7, IKBLApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-284-741