Learn More
Aviva Systems Biology Corp NFKBIL2 Antibody - middle region (P100874_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp P100874_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NFKBIL2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human NFKBIL2. Alternate names: FLJ40087, IKBR, NFKBIL2ApplicationWB. Species reactivity: Dog, Human, Mouse, Rat
Catalog No. 50-014-62756