Learn More
Aviva Systems Biology Corp Nfkbiz Antibody - C-terminal region (ARP57676_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57676_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Nfkbiz. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: AA408868, INAP, MailApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-286-346