Learn More
Proteintech Group Inc NFKBIZ Polyclonal antibody
SDP
Supplier:  Proteintech Group Inc 140141AP150UL
Encompass_Preferred
The NFKBIZ antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human NFKBIZ. This antibody recognizes human, mouse, rat antigen. The NFKBIZ antibody has been validated for the following applications: IF, WB,ELISA analysis.
Catalog No. 50-225-6391