Learn More
Cayman Chemical NformylNleLeuPheNleTyr 10mg
SDP
Catalog No. 502910603
Encompass_Preferred
A chemotactic peptide; binds to the FPR (Kd = 2.2 nM); has been conjugated to various fluorophores to monitor FPR internalization
Catalog No. 50-291-0603 Supplier Cayman Chemical Supplier No. 3586510 MG
A chemotactic peptide; binds to the FPR (Kd = 2.2 nM); has been conjugated to various fluorophores to monitor FPR internalization