Learn More
Cayman Chemical N-formyl-Nle-Leu-Phe-Nle-Tyr-Lys (trifluoroacetate salt), Purity: ≥98%, CAS: 2918767-97-0, 5 mg
SDP
Catalog No. 502912604
Encompass_Preferred
A chemotactic peptide; binds to the FPR (Kd = 2.2 nM); has been conjugated to various fluorophores to monitor FPR internalization
Catalog No. 50-291-2604 Supplier Cayman Chemical Supplier No. 358655 MG
A chemotactic peptide; binds to the FPR (Kd = 2.2 nM); has been conjugated to various fluorophores to monitor FPR internalization