Learn More
Labelmaster Inc Nfpa Diamond Only Label, 3/4" X 3/4", Paper, Roll of 1000.
SDP
Catalog No. NC9953871
Encompass_Preferred
Labelmaster's National Fire Protection Association (NFPA) Diamond Only Labels are designed for easy identification of chemical hazards by firefighters and emergency responders who are concerned with burning, spilled or exploding materials. These safety labels meet the NFPA 704 standard and are printed with numerical ratings for health hazards, fire hazards, instability and specific hazards associated with a chemical under emergency fire or spill conditions. These labels take up less space because they do not include NFPA hazard ratings or hazard warning information. The 3/4 x 3/4-in. labels are printed on pressure-sensitive, high-gloss paper and are sold in rolls of 1000.
Catalog No. NC9953871 Supplier Labelmaster Inc Supplier No. HL34
Labelmaster's National Fire Protection Association (NFPA) Diamond Only Labels are designed for easy identification of chemical hazards by firefighters and emergency responders who are concerned with burning, spilled or exploding materials. These safety labels meet the NFPA 704 standard and are printed with numerical ratings for health hazards, fire hazards, instability and specific hazards associated with a chemical under emergency fire or spill conditions. These labels take up less space because they do not include NFPA hazard ratings or hazard warning information. The 3/4 x 3/4-in. labels are printed on pressure-sensitive, high-gloss paper and are sold in rolls of 1000.