Learn More
Grainger Badger Tag and Label Corp NFPA Label, Non-Reflective, Paper, 2-7/8 in x 1-1/8 in, 500/Pk.
Catalog No. NC9574614
Encompass_Preferred
These non-reflective NFPA labels are designed for clear hazard communication and identification. Constructed from durable paper, each label features a permanent adhesive backing suitable for both indoor and outdoor applications.
  • Rectangle shape measuring 2-7/8 in. x 1-1/8 in.
  • Preprinted with fields for chemical name, common name, and manufacturer
  • Includes pictogram imagery
  • Self-laminating surface for added durability
  • Service temperature range of -4 °F to 176 °F
  • Adhesive mounting
  • Pack of 500 labels
Catalog No. NC9574614 Supplier Grainger Supplier No. 8A206
These non-reflective NFPA labels are designed for clear hazard communication and identification. Constructed from durable paper, each label features a permanent adhesive backing suitable for both indoor and outdoor applications.
  • Rectangle shape measuring 2-7/8 in. x 1-1/8 in.
  • Preprinted with fields for chemical name, common name, and manufacturer
  • Includes pictogram imagery
  • Self-laminating surface for added durability
  • Service temperature range of -4 °F to 176 °F
  • Adhesive mounting
  • Pack of 500 labels