Learn More
Cayman Chemical NFRSF7CD27 EXrAClular Do 100ug
SDP
Catalog No. 502947659
Encompass_Preferred
Source: Active recombinant C-terminal human IgG1 Fc-His-tagged CD27 expressed in HEK293 cells • Amino acids: 21-192 • MW: 47.2 kDa
Catalog No. 50-294-7659 Supplier Cayman Chemical Supplier No. 31828100 UG
Source: Active recombinant C-terminal human IgG1 Fc-His-tagged CD27 expressed in HEK293 cells • Amino acids: 21-192 • MW: 47.2 kDa