Learn More
Aviva Systems Biology Corp NFS1 Antibody - middle region (ARP33115_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33115_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against NFS1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human NFS1. Alternate names: IscS, NIFS, HUSSY-08ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-014-54888