Learn More
Aviva Systems Biology Corp NFX1 Antibody - middle region (P100680_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp P100680_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NFX1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human NFX1. Alternate names: DKFZp779G2416, MGC20369, NFX2ApplicationIHC, WB. Species reactivity: Cow, Dog, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-62679