Learn More
ABclonal Technology NFYA Rabbit mAb
SDP
Catalog No. 5000011018
Encompass_Preferred
The protein encoded by this gene is one subunit of a trimeric complex forming a highly conserved transcription factor that binds to CCAAT motifs in the promoter regions in a variety of genes Subunit A associates with a tight dimer composed of the B and C subunits resulting in a trimer that binds to DNA with high specificity and affinity The sequence specific interactions of the complex are made by the A subunit suggesting a role as the regulatory subunit In addition there is evidence of post-transcriptional regulation in this gene product either by protein degradation or control of translation Further regulation is represented by alternative splicing in the glutamine-rich activation domain with clear tissue-specific preferences for the two isoforms
Catalog No. 50-000-11018 Supplier ABclonal Technology Supplier No. A25583100UL
The protein encoded by this gene is one subunit of a trimeric complex forming a highly conserved transcription factor that binds to CCAAT motifs in the promoter regions in a variety of genes Subunit A associates with a tight dimer composed of the B and C subunits resulting in a trimer that binds to DNA with high specificity and affinity The sequence specific interactions of the complex are made by the A subunit suggesting a role as the regulatory subunit In addition there is evidence of post-transcriptional regulation in this gene product either by protein degradation or control of translation Further regulation is represented by alternative splicing in the glutamine-rich activation domain with clear tissue-specific preferences for the two isoforms