Learn More
Aviva Systems Biology Corp NFYB Antibody - N-terminal region (ARP38748_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38748_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NFYB. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human NFYB. Alternate names: HAP3, CBF-A, CBF-B, NF-YBApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-56781