Learn More
Medchemexpress LLC NG-497 | 2598242-66-9 | 99.07% | 315.36 | 1 ML
SDP
Catalog No. 5000360030
Encompass_Preferred
NG-497 is a selective human adipose triglyceride lipase (ATGL) inhibitor that targets the enzymatically active patatin-like domain of human ATGL. It has potential value for tumor research. NG-497 selectively inactivates human and nonhuman primate ATGL but not structurally and functionally related lipid hydrolases, and abolishes human adipocytes in a dose-dependent and reversible manner lipolysis. NG-497 (40 μM, 80 min) reduces lipolysis-dependent respiration in HepG2 cells without impairing mitochondrial function through off-target effects.
  • Selective human ATGL inhibitor
  • Targets enzymatically active patatin-like domain
  • Potential for tumor research
  • Selectively inactivates human and nonhuman primate ATGL
  • Abolishes human adipocytes in a dose-dependent and reversible manner lipolysis
Catalog No. 50-003-60030 Supplier Medchemexpress LLC Supplier No. HY14875610MM/1ML
NG-497 is a selective human adipose triglyceride lipase (ATGL) inhibitor that targets the enzymatically active patatin-like domain of human ATGL. It has potential value for tumor research. NG-497 selectively inactivates human and nonhuman primate ATGL but not structurally and functionally related lipid hydrolases, and abolishes human adipocytes in a dose-dependent and reversible manner lipolysis. NG-497 (40 μM, 80 min) reduces lipolysis-dependent respiration in HepG2 cells without impairing mitochondrial function through off-target effects.
  • Selective human ATGL inhibitor
  • Targets enzymatically active patatin-like domain
  • Potential for tumor research
  • Selectively inactivates human and nonhuman primate ATGL
  • Abolishes human adipocytes in a dose-dependent and reversible manner lipolysis