Learn More
Medchemexpress LLC NG-497 | 2598242-66-9 | 99.1% | 315.36 | 5 MG
SDP
Catalog No. 5000362542
Encompass_Preferred
NG-497 is a selective human adipose triglyceride lipase (ATGL) inhibitor that targets the enzymatically active patatin-like domain of human ATGL. It has potential value for tumor research. This product is for research use only.
  • Selectively inactivates human and nonhuman primate ATGL.
  • Does not inactivate structurally and functionally related lipid hydrolases.
  • Abolishes human adipocytes in a dose-dependent and reversible manner of lipolysis.
  • Reduces lipolysis-dependent respiration in HepG2 cells.
  • Does not impair mitochondrial function through off-target effects.
Catalog No. 50-003-62542 Supplier Medchemexpress LLC Supplier No. HY1487565MG
NG-497 is a selective human adipose triglyceride lipase (ATGL) inhibitor that targets the enzymatically active patatin-like domain of human ATGL. It has potential value for tumor research. This product is for research use only.
  • Selectively inactivates human and nonhuman primate ATGL.
  • Does not inactivate structurally and functionally related lipid hydrolases.
  • Abolishes human adipocytes in a dose-dependent and reversible manner of lipolysis.
  • Reduces lipolysis-dependent respiration in HepG2 cells.
  • Does not impair mitochondrial function through off-target effects.