Learn More
Selleck Chemical LLC NG 52
SDP
Catalog No. 5000002919
Encompass_Preferred
NG-52 is a tri-substituted purine that binds to the ATP-binding site of yeast cyclin-dependent kinases inhibiting Cdc28p and Pho85p with IC50s of 7 and 2 M respectively
Catalog No. 50-000-02919 Supplier Selleck Chemical LLC Supplier No. S65402MG
NG-52 is a tri-substituted purine that binds to the ATP-binding site of yeast cyclin-dependent kinases inhibiting Cdc28p and Pho85p with IC50s of 7 and 2 M respectively