Learn More
Medchemexpress LLC HY-15154 5mg Medchemexpress, NG 52 CAS:212779-48-1 Purity:>98%
SDP
Catalog No. 502081684
Encompass_Preferred
Medchemexpress, HY-15154 5mg NG 52 CAS:212779-48-1 NG 52 (Compound 52) is a potent, cell-permeable, selective, ATP-compatible and orally activeCdc28p and Pho85p kinase inhibitor with IC50s of 7 μM and 2 μM, respectively. NG 52 also inhibits the activity of phosphoglycerate kinase 1 (PGK1) with an IC50 of 2.5 μM. NG 52 is inactive against yeast kinases Kin28p, Srb10, and Cak1p. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1684 Supplier Medchemexpress LLC Supplier No. HY151545MG
Medchemexpress, HY-15154 5mg NG 52 CAS:212779-48-1 NG 52 (Compound 52) is a potent, cell-permeable, selective, ATP-compatible and orally activeCdc28p and Pho85p kinase inhibitor with IC50s of 7 μM and 2 μM, respectively. NG 52 also inhibits the activity of phosphoglycerate kinase 1 (PGK1) with an IC50 of 2.5 μM. NG 52 is inactive against yeast kinases Kin28p, Srb10, and Cak1p. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.