Learn More
Enzo Life Sciences NG-Hydroxy-L-arginine . monoacetate (25mg). CAS: 53598-01-9
SDP
Catalog No. 502007735
Encompass_Preferred
Intermediate in NO biosynthesis. Intermediate in the biosynthesis of nitric oxide from L-arginine by nitric oxide synthases (NOS). Inhibitor of arginases. Alternative name: L-HOArg . AcOH. Formula: C6H14N4O3 . CH3COOH. MW: 190.2 . 60.1. Purity: ≥98% (HPLC). Appearance: White to off-white solid. Solubility: Soluble in water (50mg/ml). Long Term Storage: +4°C.
Catalog No. 50-200-7735 Supplier Enzo Life Sciences Supplier No. ALX106004M025
Intermediate in NO biosynthesis. Intermediate in the biosynthesis of nitric oxide from L-arginine by nitric oxide synthases (NOS). Inhibitor of arginases. Alternative name: L-HOArg . AcOH. Formula: C6H14N4O3 . CH3COOH. MW: 190.2 . 60.1. Purity: ≥98% (HPLC). Appearance: White to off-white solid. Solubility: Soluble in water (50mg/ml). Long Term Storage: +4°C.